3-Boc-3,8-diazabicyclo[4.2.0]octane 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3,8-diazabicyclo[4.2.0]octane-3-carboxylate

OR317211-250MG
APOSOR317211-250MGEA 410 EUR
APOSOR317211-250MG
3-Boc-3,8-diazabicyclo[4.2.0]octane 97%
3-Boc-3,8-diazabicyclo[4.2.0]octane
Formel: C₁₁H₂₀N₂O₂
Molecular Weight: 212,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056864
CAS-Nummer: 928754-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR