1-Boc-2-methylazetidine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR317213-1G
APOSOR317213-1GEA
463
EUR
APOSOR317213-1G
1-Boc-2-methylazetidine-2-carboxylic acid
1-Boc-2-methylazetidine-2-carboxylic acid
Formel:
C₁₀H₁₇NO₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD12406956 CAS-Nummer: 449758-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...