Methyl-2-azetidinecarboxylate hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl azetidine-2-carboxylate hydrochloride
OR317227-1G
OR317227-5G
APOSOR317227-1GEA
115
EUR
APOSOR317227-1G
APOSOR317227-5G
Methyl-2-azetidinecarboxylate hydrochloride 95%
Methyl-2-azetidinecarboxylate hydrochloride
Formel:
C₅H₁₀ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861757 CAS-Nummer: 162698-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...