N,N,3-Trimethylazetidin-3-amine hydrochloride

Lieferant: Apollo Scientific

OR317229-1G
APOSOR317229-1GEA 540 EUR
APOSOR317229-1G
N,N,3-Trimethylazetidin-3-amine hydrochloride
N,N,3-Trimethylazetidin-3-amine hydrochloride
Formel: C₆H₁₅ClN₂
Molecular Weight: 150.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123304
CAS-Nummer: 132771-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR