3-[(tert-Butyldimethylsilanyl)oxy]azetidine

Lieferant: Apollo Scientific
Achtung

OR317233-1G OR317233-250MG OR317233-5G
APOSOR317233-1GEA 364 EUR
APOSOR317233-1G APOSOR317233-250MG APOSOR317233-5G
3-[(tert-Butyldimethylsilanyl)oxy]azetidine
3-[(tert-Butyldimethylsilanyl)oxy]azetidine
Formel: C₉H₂₁NOSi
Molecular Weight: 187.36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20662828
CAS-Nummer: 875340-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR