3-Cyano-1-azetidinesulphonylchloride

Lieferant: Apollo Scientific
Gefahr

OR317236-1G
APOSOR317236-1GEA 429 EUR
APOSOR317236-1G
3-Cyano-1-azetidinesulphonylchloride
3-Cyano-1-azetidinesulphonylchloride
Formel: C₄H₅ClN₂O₂S
Molecular Weight: 180,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22421688
CAS-Nummer: 1310734-08-7
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR