2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Oxa-6-azaspiro[3.3]heptane hemioxalate
OR317237-1G
APOSOR317237-1GEA
21.9
EUR
APOSOR317237-1G
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) 97%
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1)
Formel:
C₁₂H₂₀N₂O₆ Molecular Weight: 144,15 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16652332 CAS-Nummer: 1045709-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...