(R)-Azetidine-2-carboxylic acid

Lieferant: Apollo Scientific

OR317238-1G
APOSOR317238-1GEA 0 EUR
APOSOR317238-1G
(R)-Azetidine-2-carboxylic acid
(R)-Azetidine-2-carboxylic acid
Formel: C₄H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00800731
CAS-Nummer: 7729-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR