7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)

Lieferant: Apollo Scientific

OR317239-1G
APOSOR317239-1GEA 0 EUR
APOSOR317239-1G
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)
Formel: C₁₆H₂₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD22422285
CAS-Nummer: 1408076-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR