(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester 97%

Lieferant: Apollo Scientific
Achtung

OR317245-1G OR317245-250MG
APOSOR317245-1GEA 0 EUR
APOSOR317245-1G APOSOR317245-250MG
(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester 97%
(R)-1-Boc-Azetidine-2-carboxylic acid methyl ester
Formel: C₁₀H₁₇NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD11042210
CAS-Nummer: 1260593-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR