Methyl-1-boc-azetidine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-Boc-azetidine-2-carboxylate, 1-Boc-Azetidine-2-carboxylic acid methyl ester

OR317246-5G
APOSOR317246-5GEA 87 EUR
APOSOR317246-5G
Methyl-1-boc-azetidine-2-carboxylate
Methyl-1-boc-azetidine-2-carboxylate
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215,2463 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657099
CAS-Nummer: 255882-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR