(S)-2-Aminomethyl-1-boc-azetidine

Lieferant: Apollo Scientific
Achtung

OR317248-1G
APOSOR317248-1GEA 473 EUR
APOSOR317248-1G
(S)-2-Aminomethyl-1-boc-azetidine
(S)-2-Aminomethyl-1-boc-azetidine
Formel: C₉H₁₈N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD17215571
CAS-Nummer: 1007873-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR