4-Chlor-7-[(4-methylphenyl)sulfonyl]-7H-pyrrolo[2,3-d]pyrimidin

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine, 4-Chloro-7-(p-toluenesulphonyl)-7H-pyrrolo[2,3-d]pyrimidine

OR317249-10G OR317249-1G OR317249-5G
APOSOR317249-10GEA 574 EUR
APOSOR317249-10G APOSOR317249-1G APOSOR317249-5G
4-Chlor-7-[(4-methylphenyl)sulfonyl]-7H-pyrrolo[2,3-d]pyrimidin
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₁₃H₁₀ClN₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09907939
CAS-Nummer: 479633-63-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR