4-Bromo-1-(oxetan-3-yl)-1H-pyrazole 97%
Lieferant: Apollo Scientific
OR317259-1G
APOSOR317259-1GEA
145
EUR
APOSOR317259-1G
4-Bromo-1-(oxetan-3-yl)-1H-pyrazole 97%
4-Bromo-1-(oxetan-3-yl)-1H-pyrazole
Formel:
C₆H₇BrN₂O Molecular Weight: 203,037 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21849011 CAS-Nummer: 1374657-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...