2-Boc-6-hydroxy-2-azaspiro[3.4]octane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-2-propanyl 6-hydroxy-2-azaspiro[3.4]octane-2-carboxylate, tert-Butyl 6-hydroxy-2-azaspiro[3.4]octane-2-carboxylate

OR317311-1G
APOSOR317311-1GEA 98.6 EUR
APOSOR317311-1G
2-Boc-6-hydroxy-2-azaspiro[3.4]octane
2-Boc-6-hydroxy-2-azaspiro[3.4]octane
Formel: C₁₂H₂₁NO₃
Molecular Weight: 227,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22415223
CAS-Nummer: 1363381-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR