3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

OR317323-1G
APOSOR317323-1GEA 82.8 EUR
APOSOR317323-1G
3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18382389
CAS-Nummer: 1330055-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR