4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Lieferant: Apollo Scientific

Synonyme: 4-chloro-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

OR317326-1G
APOSOR317326-1GEA 105 EUR
APOSOR317326-1G
4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
4-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅ClN₂O
Molecular Weight: 168,5804 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11518939
CAS-Nummer: 346599-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR