5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Lieferant: Apollo Scientific

OR317327-1G
APOSOR317327-1GEA 337 EUR
APOSOR317327-1G
5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12962791
CAS-Nummer: 1190314-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR