5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
Lieferant: Apollo Scientific
Synonyme:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
OR317332-1G
APOSOR317332-1GEA
789
EUR
APOSOR317332-1G
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
Formel:
C₁₃H₁₇BN₂O₃ Molecular Weight: 260,097 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16995984 CAS-Nummer: 1207623-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...