4-tert-Butoxybenzoic acid 95+%

Lieferant: Apollo Scientific

OR318017-1G OR318017-25G OR318017-5G
APOSOR318017-1GEA 88.6 EUR
APOSOR318017-1G APOSOR318017-25G APOSOR318017-5G
4-tert-Butoxybenzoic acid 95+%
4-tert-Butoxybenzoic acid
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00082602
CAS-Nummer: 13205-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR