tert-Butyl ethyl(piperidin-4-yl)carbamate 95+%

Lieferant: Apollo Scientific
Achtung

OR318029-1G
APOSOR318029-1GEA 335 EUR
APOSOR318029-1G
tert-Butyl ethyl(piperidin-4-yl)carbamate 95+%
tert-Butyl ethyl(piperidin-4-yl)carbamate
Formel: C₁₂H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06808579
CAS-Nummer: 313977-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR