4-(Methoxycarbonyl)nicotinic acid 90%

Lieferant: Apollo Scientific
Achtung

OR318041-1G
APOSOR318041-1GEA 232 EUR
APOSOR318041-1G
4-(Methoxycarbonyl)nicotinic acid 90%
4-(Methoxycarbonyl)nicotinic acid
Formel: C₈H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00111513
CAS-Nummer: 24202-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR