Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate 95+%

Lieferant: Apollo Scientific

OR318043-1G
APOSOR318043-1GEA 359 EUR
APOSOR318043-1G
Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate 95+%
Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Formel: C₇H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD10688281
CAS-Nummer: 33458-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR