2-tert-Butyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95+%

Lieferant: Apollo Scientific

OR318046-1G
APOSOR318046-1GEA 630 EUR
APOSOR318046-1G
2-tert-Butyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95+%
2-tert-Butyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Formel: C₁₂H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11052308
CAS-Nummer: 1049676-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR