2-Amino-5-methylnicotinaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-methylpyridine-3-carboxaldehyde

OR318050-1G
APOSOR318050-1GEA 957 EUR
APOSOR318050-1G
2-Amino-5-methylnicotinaldehyde 95+%
2-Amino-5-methylnicotinaldehyde
Formel: C₇H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09754122
CAS-Nummer: 1023814-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR