4-Benzyl-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95+%

Lieferant: Apollo Scientific

OR318051-1G
APOSOR318051-1GEA 1440 EUR
APOSOR318051-1G
4-Benzyl-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95+%
4-Benzyl-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid
Formel: C₁₇H₂₄N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09026073
CAS-Nummer: 1214057-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR