1-Phenethylguanidine hemisulphate 95+%

Lieferant: Apollo Scientific

OR318052-1G
APOSOR318052-1GEA 685 EUR
APOSOR318052-1G
1-Phenethylguanidine hemisulphate 95+%
1-Phenethylguanidine hemisulphate
Formel: 2C₉H₁₃N₃·H₂SO₄
Molecular Weight: 424,518 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035581
CAS-Nummer: 2551-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR