2-(4-Acetylpiperazin-1-yl)ethanamine 95+%

Lieferant: Apollo Scientific

OR318063-1G
APOSOR318063-1GEA 696 EUR
APOSOR318063-1G
2-(4-Acetylpiperazin-1-yl)ethanamine 95+%
2-(4-Acetylpiperazin-1-yl)ethanamine
Formel: C₈H₁₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD08275715
CAS-Nummer: 148716-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR