tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR318070-5G OR318070-10G
APOSOR318070-5GEA 37.7 EUR
APOSOR318070-5G APOSOR318070-10G
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl-3-methyl-4-oxopiperidine-1-carboxylate
Formel: C₁₁H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08460962
CAS-Nummer: 181269-69-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR