5-Methyl-6-morpholinopyridin-3-ylboronic acid hydrochloride 95+%

Lieferant: Apollo Scientific

OR318075-1G
APOSOR318075-1GEA 807 EUR
APOSOR318075-1G
5-Methyl-6-morpholinopyridin-3-ylboronic acid hydrochloride 95+%
5-Methyl-6-morpholinopyridin-3-ylboronic acid hydrochloride
Formel: C₁₀H₁₆BClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11040540
CAS-Nummer: 1231955-78-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR