1,6-Naphthyridin-5(6H)-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR318077-250MG
APOSOR318077-250MGEA
109
EUR
APOSOR318077-250MG
1,6-Naphthyridin-5(6H)-one 97%
1,6-Naphthyridin-5(6H)-one
Formel:
C₈H₆N₂O Schmelzpunkt: 239…241 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08567947 CAS-Nummer: 23616-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...