1,6-Naphthyridine-5-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR318079-1G
APOSOR318079-1GEA 1210 EUR
APOSOR318079-1G
1,6-Naphthyridine-5-carboxylic acid 95+%
1,6-Naphthyridine-5-carboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174.16 g/mol
Schmelzpunkt: 193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06762718
CAS-Nummer: 74048-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR