8-Bromo-1,6-naphthyridin-5(6H)-one

Lieferant: Apollo Scientific
Achtung

OR318080-1G
APOSOR318080-1GEA 290 EUR
APOSOR318080-1G
8-Bromo-1,6-naphthyridin-5(6H)-one
8-Bromo-1,6-naphthyridin-5(6H)-one
Formel: C₈H₅BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00928592
CAS-Nummer: 155057-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR