8-(Piperazin-1-yl)-1,2,3,4-tetrahydro-1,7-naphthyridine dihydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

OR318085-1G
APOSOR318085-1GEA 1110 EUR
APOSOR318085-1G
8-(Piperazin-1-yl)-1,2,3,4-tetrahydro-1,7-naphthyridine dihydrochloride 95+%
8-(Piperazin-1-yl)-1,2,3,4-tetrahydro-1,7-naphthyridine dihydrochloride
Formel: C₁₂H₂₀Cl₂N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11052318
CAS-Nummer: 1233513-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR