2,3-Diphenyl-1,4,8-triazaspiro[4.5]deca-1,3-diene 95+%

Lieferant: Apollo Scientific

OR318087-1G
APOSOR318087-1GEA 411 EUR
APOSOR318087-1G
2,3-Diphenyl-1,4,8-triazaspiro[4.5]deca-1,3-diene 95+%
2,3-Diphenyl-1,4,8-triazaspiro[4.5]deca-1,3-diene
Formel: C₁₉H₁₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11052319
CAS-Nummer: 1049676-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR