3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%

Lieferant: Apollo Scientific

OR318107-1G
APOSOR318107-1GEA 957 EUR
APOSOR318107-1G
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine
Formel: C₁₂H₁₂BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258796
CAS-Nummer: 916423-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR