3-(3-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%

Lieferant: Apollo Scientific
Achtung

OR318108-1G
APOSOR318108-1GEA 957 EUR
APOSOR318108-1G
3-(3-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(3-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine
Formel: C₁₂H₁₂BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258797
CAS-Nummer: 916423-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR