3-(4-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Lieferant: Apollo Scientific
OR318109-1G
APOSOR318109-1GEA
957
EUR
APOSOR318109-1G
3-(4-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(4-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine
Formel:
C₁₂H₁₂BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258798 CAS-Nummer: 916423-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...