2-Methyl-5-(methylsulphonyl)aniline 95+%
Lieferant: Apollo Scientific
OR318114-1G
APOSOR318114-1GEA
0
EUR
APOSOR318114-1G
2-Methyl-5-(methylsulphonyl)aniline 95+%
2-Methyl-5-(methylsulphonyl)aniline
Formel:
C₈H₁₁NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258802 CAS-Nummer: 1671-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...