5-Bromo-1H-indol-7-amine 95+%

Lieferant: Apollo Scientific

OR318115-1G
APOSOR318115-1GEA 0 EUR
APOSOR318115-1G
5-Bromo-1H-indol-7-amine 95+%
5-Bromo-1H-indol-7-amine
Formel: C₈H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02093961
CAS-Nummer: 374537-99-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR