2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide 95+%

Lieferant: Apollo Scientific

OR318128-1G
APOSOR318128-1GEA 1220 EUR
APOSOR318128-1G
2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide 95+%
2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide
Formel: C₈H₉NO₃S
Molecular Weight: 199,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28103339
CAS-Nummer: 156359-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR