5-Aminopyridine-3-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Aminopyridine-5-boronic acid, pinacol ester, 3-Aminopyridine-5-boronic acid pinacol ester

OR3184-1G
APOSOR3184-1GEA 60.9 EUR
APOSOR3184-1G
5-Aminopyridine-3-boronic acid, pinacol ester
5-Aminopyridine-3-boronic acid, pinacol ester
Formel: C₁₁H₁₇BN₂O₂
Molecular Weight: 220,076 g/mol
Schmelzpunkt: 194…195 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09260489
CAS-Nummer: 1073354-99-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR