4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 4,4-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-enone

OR3187-100MG OR3187-50MG
APOSOR3187-100MGEA 55.1 EUR
APOSOR3187-100MG APOSOR3187-50MG
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronsäurepinakolester
Formel: C₁₄H₂₃BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09027074
CAS-Nummer: 219489-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR