2-Methyl-5-nitrophenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-5-nitrophenylboronic acid, pinacol ester, 2-Methyl-5-nitrophenylboronic Acid Pinacol Ester

OR3190-1G
APOSOR3190-1GEA 109 EUR
APOSOR3190-1G
2-Methyl-5-nitrophenylboronsäurepinakolester 98%
2-Methyl-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₄
Molecular Weight: 263,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027081
CAS-Nummer: 957062-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR