4-Acetyl-1-phenyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1-Phenyl-1H-pyrazol-4-yl)ethanone

OR3198-1G
APOSOR3198-1GEA 81.2 EUR
APOSOR3198-1G
4-Acetyl-1-phenyl-1H-pyrazole
4-Acetyl-1-phenyl-1H-pyrazole
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159631
CAS-Nummer: 3968-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR