4-[4-(Pyridin-2-yl)-1H-pyrazol-3-yl]morpholine 95%

Lieferant: Apollo Scientific

OR32001-1G
APOSOR32001-1GEA 1570 EUR
APOSOR32001-1G
4-[4-(Pyridin-2-yl)-1H-pyrazol-3-yl]morpholine 95%
4-[4-(Pyridin-2-yl)-1H-pyrazol-3-yl]morpholine
Formel: C₁₂H₁₄N₄O
Molecular Weight: 230,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127011
CAS-Nummer: 240115-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR