5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR32002-1G
APOSOR32002-1GEA 1570 EUR
APOSOR32002-1G
5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile 95%
5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile
Formel: C₁₁H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00172106
CAS-Nummer: 24400-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR