4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one 95%

Lieferant: Apollo Scientific

OR32003-1G
APOSOR32003-1GEA 1570 EUR
APOSOR32003-1G
4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one 95%
4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one
Formel: C₈H₁₃N₃O₂
Molecular Weight: 183,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138266
CAS-Nummer: 240799-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR