o-Tolylmagnesium chloride 1 M in THF

Lieferant: Apollo Scientific

OR320032-100ML
APOSOR320032-100MLEA 145 EUR
APOSOR320032-100ML
o-Tolylmagnesium chloride 1 M in THF
o-Tolylmagnesium chloride
Formel: C₇H₇ClMg
Molecular Weight: 150,888 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 33872-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR