5-[(E)-2-(Dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR32005-1G
APOSOR32005-1GEA 1570 EUR
APOSOR32005-1G
5-[(E)-2-(Dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carbonitrile 95%
5-[(E)-2-(Dimethylamino)ethenyl]-3-phenyl-1,2-oxazole-4-carbonitrile
Formel: C₁₄H₁₃N₃O
Molecular Weight: 239,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127045
CAS-Nummer: 1610688-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR